missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Biotin Ab-2 Mouse Monoclonal Antibody, 200μg/mL with BSA and azide

Detect both the free and protein-conjugated forms of biotin with Epredia™ Biotin Ab-2, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-1048-P1ABX

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12648706

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.


Description

Description

Ab-1 has a very high affinity for biotin and is excellent for use in various amplification techniques. It recognizes both the free and protein-conjugated (either soluble or cell bound) form of biotin. In some applications, localization of biotinylated probes with avidin produces unacceptably high background staining. Anti-biotin antibody may be substituted to decrease this noise.

Host Species: Mouse

Clone: BTN.4

Isotype: IgG1/κ

Species Reactivity: All species.

Immunogen: Biotin conjugated to BSA

Recommended for:

  • Western Blotting
  • Confocal Microscopy
  • Flow Cytometry
  • Image Analysis
  • In situ Nucleic Acid Hybridization

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Biotin Ab-2
Monoclonal
1mg/mL
Mouse
100 μL
Epitope Tags
All species
IgG2a κ
Western Blot
BTN.4
Unconjugated
Biotin conjugated to BSA
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions