missing translation for 'onlineSavingsMsg'
Learn More

CD155/PVR (Poliovirus Receptor) Ab-1 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunofluorescence and flow cytometry applications.

Brand:  Epredia MS-465-P1

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12661517

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.


Description

Description

PVR is a member of Ig-superfamily with 3 Ig-domains in arrangement V-C-C. Four mRNAs are produced from a single gene for PVR, (mapped to human chromosome band 19q13.1-13.2). Two of the mRNAs (called H20A and H20B or α and κ,) encode cell surface molecules of about 43kDa and 45kDa polypeptide backbones, respectively. Two other splice variants, Β and γ, encode secreted molecules of about 44kDa. The cytoplasmic tails are short (aa 35-50) and are rich in serine phosphorylated residues.

Host Species: Mouse

Clone: D171

Isotype: IgG1

Species Reactivity: Human and Monkey. Does not react with pig, dog, rabbit, rat, hamster and mouse. Others not known.

Epitope: aa 35-50

Immunogen: HeLa cells

Molecular Weight: 60-80kDa

Positive Control: HeLa, SV 80, HEp-2, RD, A431, HL-60, U-937 cells; Monkey Vero, GMK, or TC-7 cells

Cellular Localization: Membrane

Recommended for:

  • Western Blotting (Not suitable)
  • Immunofluorescence
  • Flow Cytometry
  • Inhibits Replication of Poliovirus in HeLa Cells

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

CD155 Ab-1
Monoclonal
200μg/mL
Mouse
500 μL
Cell Adhesion
Human, Non-human Primate
IgG1
Immunofluorescence
D171
Unconjugated
HeLa cells
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions