missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD24 (GPI-Linked Surface Mucin) Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Ensure accurate, reproducible results in immunohistochemistry, western blotting, and flow cytometry experiments with Epredia™ CD24 (GPI-Linked Surface Mucin) Ab-1.

Brand:  Epredia™ MS-1278-P0

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12659446

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.


Description

Description

CD24 is a two-chain glycosylphosphatidylinositol (GPI)-anchored glycoprotein expressed at multiple stages of B-cell development, beginning with the bone marrow pro-B-cell compartment and continuing through mature, surface Ig positive B-cells. Plasma cell expression is very low or negative. It is also expressed on the majority of B-lineage acute lymphoblastic leukemias, B-cell CCLs and B-cell non-Hodgkin's lymphomas. CD24 may play a role in regulation of B-cell proliferation and maturation.

Host Species: Mouse

Clone: SN3

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Cell membrane antigen preparation isolated from NALM-1 human pre-B leukemia cell line

Molecular Weight: 30-70 kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Frozen only)
  • Western Blotting
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

CD24 (GPI-Linked Surface Mucin) Ab-1
Monoclonal
200μg/mL
Mouse
100 μL
B-Cell
Human
IgG2a κ
Flow Cytometry, Immunohistochemistry (Frozen), Western Blot
SN3
Unconjugated
Cell membrane antigen preparation isolated from NALM-1 human pre-B leukemia cell line
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions