Learn More
CD68/Macrophage Marker Ab-3 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD68/Macrophage Marker Ab-3, Mouse Monoclonal Antibody.
Brand: Epredia MS-397-P1ABX
Additional Details : Weight : 0.00010kg
Description
CD68 is expressed on macrophages and monocytes.Host Species: Mouse
Clone: KP1
Isotype: IgG1/κ
Species Reactivity: Human, Monkey, Cat and Rat. Does not react with pig, dog and chicken.
Epitope: Not determined
Immunogen: Subcellular fraction of human alveolar macrophages
Molecular Weight: 110 kDa
Positive Control: Tonsil, lymph node, or spleen
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
CD68/Macrophage Marker Ab-3 | |
Monoclonal | |
1mg/mL | |
Mouse | |
100 μL | |
Hematopoietic | |
Feline, Human, Non-human Primate, Rat | |
IgG1 κ |
Immunohistochemistry (Paraffin) | |
KP1 | |
Unconjugated | |
Subcellular fraction of human alveolar macrophages | |
IVD | |
Primary | |
Concentrated |