missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD8 Rabbit Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry applications with Epredia™ CD8, Rabbit Monoclonal Antibody.

Brand:  Epredia™ RM-9116-RQ

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 12673737

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.


Description

Description

CD8 molecule consists of two chains, termed α and β chain, which are expressed as a disulphide-linked α/β heterdimer or as an α/α homodimer on T cell subset, thymocytes and NK cells. The majority of CD8+ T cells express CD8 as α/β heterdimer. CD8 functions as a coreceptor in concert with TCR for binding the MHC class I/peptide complex. The HIV-2 envelope glycoprotein binds CD8 alpha chain (but not β chain).

Host Species: Rabbit

Clone: SP16

Species Reactivity: Human. Others not tested.

Epitope: C-terminus

Immunogen: Synthetic peptide sequence comprising the 13 C-terminal amino acids of the cytoplasmic domain of α-chain of the CD8 molecule

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

CD8
Monoclonal
Unconjugated
Synthetic peptide sequence comprising the 13 C-terminal amino acids of the cytoplasmic domain of alpha chain of the CD8 molecule
IVD
Primary
Immunohistochemistry (Paraffin)
SP16
Rabbit
12 mL
T-Cell
Ready-to-Use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions