missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ CD8 Rabbit Monoclonal Antibody
Ensure accurate, reproducible results in immunohistochemistry applications with Epredia™ CD8, Rabbit Monoclonal Antibody.
165.00€ - 790.00€
Specifications
Antigen | CD8 |
---|---|
Clone | SP16 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Host Species | Rabbit |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12623747
|
Epredia™
RM-9116-S1 |
Concentrated | 500 μL |
434.00€
500µL |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12603747
|
Epredia™
RM-9116-S |
Concentrated | 1 mL |
790.00€
1mL |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12683737
|
Epredia™
RM-9116-R7 |
Prediluted | 7 mL |
369.00€
7mL |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12613747
|
Epredia™
RM-9116-S0 |
Concentrated | 100 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
12673737
|
Epredia™
RM-9116-RQ |
Ready-to-Use | 12 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
Description
CD8 molecule consists of two chains, termed α and β chain, which are expressed as a disulphide-linked α/β heterdimer or as an α/α homodimer on T cell subset, thymocytes and NK cells. The majority of CD8+ T cells express CD8 as α/β heterdimer. CD8 functions as a coreceptor in concert with TCR for binding the MHC class I/peptide complex. The HIV-2 envelope glycoprotein binds CD8 alpha chain (but not β chain).Host Species: Rabbit
Clone: SP16
Species Reactivity: Human. Others not tested.
Epitope: C-terminus
Immunogen: Synthetic peptide sequence comprising the 13 C-terminal amino acids of the cytoplasmic domain of α-chain of the CD8 molecule
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
CD8 | |
Immunohistochemistry (Paraffin) | |
Rabbit | |
T-Cell | |
Primary |
SP16 | |
Monoclonal | |
IVD | |
Synthetic peptide sequence comprising the 13 C-terminal amino acids of the cytoplasmic domain of alpha chain of the CD8 molecule |