missing translation for 'onlineSavingsMsg'
Learn More

Chromogranin A Ab-3 Mouse Monoclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Chromogranin A Ab-3, Mouse Monoclonal Antibody.

132.00€ - 416.00€

Specifications

Antigen Chromogranin A Ab-3
Clone LK2H10 + PHE5
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 6
Product Code Brand Form Quantity Price Quantity & Availability  
Product Code Brand Form Quantity Price Quantity & Availability  
12651267
View Documents Promotion Details
Epredia
MS-382-P1
Concentrated 500 μL
278.00€
500µL
View alternative products
Save Up to 
»
Estimated Shipment: 04-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
12631267
View Documents Promotion Details
Epredia
MS-382-P
Concentrated 1 mL
390.00€
1mL
View alternative products
Save Up to 
»
Estimated Shipment: 04-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
12671267
View Documents Promotion Details
Epredia
MS-382-PABX
Concentrated 200 μL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
12641267
View Documents Promotion Details
Epredia
MS-382-P0
Concentrated 100 μL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
12661267
View Documents Promotion Details
Epredia
MS-382-P1ABX
Concentrated 100 μL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
12601277
View Documents Promotion Details
Epredia
MS-382-R7
Prediluted 7 mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
Description

Description

Chromogranin A (a protein of 439-amino acid which is encoded on chromosome 14) is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, pheo-chromocytomas, paragangliomas, and other neuro-endocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms. Ab-3 cocktail is especially designed for sensitive detection of Chromogranin A in formalin-fixed, paraffin-embedded tissues.

Host Species: Mouse

Clone: LK2H10 + PHE5

Isotype: IgG1 + IgG1

Species Reactivity: Human and Monkey. Others not known.

Immunogen: Human pheochromocytoma

Molecular Weight: 68-75 kDa

Positive Control: Pancreas

Cellular Localization: Cytoplasmic in endocrine cells

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Chromogranin A Ab-3
Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Non-human Primate
IgG1
LK2H10 + PHE5
Monoclonal
Mouse
Oncogenesis
Human pheochromocytoma
Primary
Videos
SDS
Documents

Documents

Product Certifications