Learn More
Epredia™ Lab Vision™ Chromogranin A Rabbit Monoclonal Antibody
Obtain accurate and reproducible results in immunohistochemistry applications with Epredia™ Chromogranin A, Rabbit Monoclonal Antibody.
Brand: Epredia™ RM-9112-S0
Additional Details : Weight : 0.00170kg
Description
Chromogranin A (a protein of 439 amino acid which is encoded on chromosome 14) is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, phenochromocytomas, paragangliomas, and other neuroendocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms.Host Species: Rabbit
Clone: SP12
Isotype: IgG
Species Reactivity: Human. Others not known
Epitope: Not determined
Immunogen: Recombinant protein encoding human chromogranin A
Positive Control: Pancreas
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Chromogranin A | |
Monoclonal | |
Unconjugated | |
Recombinant protein encoding human chromogranin A | |
IVD | |
Primary | |
Concentrated |
Immunohistochemistry (Paraffin) | |
SP12 | |
Rabbit | |
100 μL | |
Oncogenesis | |
Human | |
IgG |