missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin D1 Rabbit Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Cyclin D1, Rabbit Monoclonal Antibody.
274.38€ - 1515.00€
Specifications
Antigen | Cyclin D1 |
---|---|
Clone | EPR2241IHC |
Applications | Immunohistochemistry |
Conjugate | Unconjugated |
Host Species | Rabbit |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12663647
|
Epredia
RM-2113-R7 |
Ready-to-Use | 7 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
12693647
|
Epredia
RM-2113-S0 |
Supernatant | 100 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
12603657
|
Epredia
RM-2113-S |
Supernatant | 1 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
Description
Cyclin D1 or PRAD-1 or bcl-1 is a key cell cycle regulator. It functions in association with cdk4 and/or cdk6 by phosphorylating the retinoblastoma protein that releases transcription factors to initiate DNA replication. It is a putative proto-oncogene overexpressed in a wide variety of human neoplasms. In lymphoid lesions, with the exception of few rare diseases, it is specific for mantle cell lymphoma (MCL).Host Species: Rabbit
Clone: EPR2241IHC
Isotype: IgG
Species Reactivity: Human. Others not tested.
Immunogen: A synthetic peptide corresponding to residues near the C-terminus of human Cyclin D1
Molecular Weight: 36 kDa
Positive Control: Mantle cell lymphoma
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Cyclin D1 | |
Immunohistochemistry | |
Rabbit | |
Primary |
EPR2241IHC | |
Unconjugated | |
A synthetic peptide corresponding to residues near the C-terminus of human Cyclin D1 |