missing translation for 'onlineSavingsMsg'
Learn More

Heat Shock Protein 90α/hsp90α (HSP86) Ab-1 Rabbit Polyclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and immunoprecipitation experiments.

Brand:  Epredia RB-119-P0

Additional Details : Weight : 0.00170kg

 View more versions of this product

Product Code. 12642837

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.


Description

Description

Cells respond to heat and chemical stress by inducing the synthesis of a group of proteins referred to as heat shock proteins. HSP90 is present in cells at high levels in the cytoplasm and is composed of two separate gene products, HSP90α and HSP90Β.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: aa 2-12

Immunogen: A synthetic 11-mer peptide, corresponding to aa 2-12 (PEETQTQDQPM-Cys) from the N-terminus end of mouse heat shock protein 90α (hsp90α)

Molecular Weight: 86 kDa

Positive Control: LS174T cells, MAD109 cells or breast CA

Cellular Localization: Cytoplasmic and nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Heat Shock Protein 90alpha/hsp90alpha (HSP86) Ab-1
Polyclonal
Unconjugated
A synthetic 11-mer peptide, corresponding to aa 2-12 (PEETQTQDQPM-Cys) from the N-terminus end of mouse heat shock protein 90alpha (hsp90alpha)
RUO
Primary
Concentrated
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
1mg/mL
Rabbit
100 μL
Stress Response
Human, Mouse
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions