Learn More
Epredia™ Lab Vision™ Hepatitis B Virus Surface Antigen (HBVsAg) Ab-1 Mouse Monoclonal Antibody
Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Hepatitis B Virus Surface Antigen (HBVsAg) Ab-1, Mouse Monoclonal Antibody.
Brand: Epredia™ MS-314-R7
Additional Details : Weight : 0.01200kg
Description
Hepatitis B virus causes type B hepatitis and it belongs to hepadnavirus family. It is spherical in shape with a diameter of 42nm. It contains a 27nm partially double stranded DNA core enclosed within a lipoprotein coat. The antigenic activity of the nucleocapsid core is designated as hepatitis B core antigen. The antigens in the outer surface are called as hepatitis B virus surface antigens. Core antigens are localized within the nuclei whereas the surface antigens are present in the cytoplasm of the infected cells. Antibodies to surface antigens appear in circulation at an early stage of infection whereas the antibodies to the core antigens are detected after several weeks.Host Species: Mouse
Clone: T9; same as 3E7
Isotype: IgG1/λ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Purified HBVsAg, subtype ayw3
Positive Control: Hepatitis B virus infected liver
Cellular Localization: Cytoplasm
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Hepatitis B Virus Surface Antigen (HBVsAg) Ab-1 | |
Monoclonal | |
Unconjugated | |
Purified HbsAg, subtype ayw3 | |
ASR | |
Primary | |
Prediluted |
Immunohistochemistry (Paraffin) | |
T9; same as 3E7 | |
Mouse | |
7 mL | |
Infectious Disease | |
Human | |
IgG1 λ |