missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin Receptor (α-subunit) Ab-2 Mouse Monoclonal Antibody, Epredia™
Epredia™ Insulin Receptor (α-Subunit) Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.
143.82€ - 846.60€
Specifications
Antigen | Insulin Receptor (alpha-Subunit) Ab-2 |
---|---|
Clone | 83-7 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Host Species | Mouse |
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
12641837
|
Epredia
MS-632-B0 |
Concentrated | Biotin | 100 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12651837
|
Epredia
MS-632-B1 |
Concentrated | Biotin | 500 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12631837
|
Epredia
MS-632-B |
Concentrated | Biotin | 1 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12671837
|
Epredia
MS-632-P0 |
Concentrated | Unconjugated | 100 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12691837
|
Epredia
MS-632-P1ABX |
Concentrated | Unconjugated | 100 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12601847
|
Epredia
MS-632-PABX |
Concentrated | Unconjugated | 200 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12664269
|
Epredia
MS-632-P1 |
Concentrated | Unconjugated | 500 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12661837
|
Epredia
MS-632-P |
Concentrated | Unconjugated | 1 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12631847
|
Epredia
MS-632-R7 |
Prediluted | Unconjugated | 7 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
Description
The human insulin receptor is a heterotetrameric membrane glycoprotein consisting of disulfide-linked subunits in a Β-α-α-Β configuration. The Β-subunit (95kDa) possesses a single transmembrane domain, whereas the α-subunit (135kDa) is completely extracellular.Host Species: Mouse
Clone: 83-7
Isotype: IgG1/κ
Species Reactivity: Human, Cow, Pig, Sheep and Rabbit. Does not react with mouse and rat.
Epitope: Exon 3
Immunogen: IM-9 lymphocytes followed by IR
Molecular Weight: 135kDa
Positive Control: IM-9 lymphocyte cells, Placenta
Cellular Localization: Cell membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- ELISA
- Flow Cytometry
- Insulin-like Agonist
- Does Not Significantly Inhibit Insulin Binding
- Tyrosine Kinase Assay (Ab-mediated capture on microplates)
Note:
USA: RUO; Int'l: RUO
Specifications
Insulin Receptor (alpha-Subunit) Ab-2 | |
Immunohistochemistry (Paraffin) | |
Mouse | |
Steroids Hormones and Growth Factors | |
IM-9 lymphocytes followed by IR | |
Primary |
83-7 | |
Monoclonal | |
RUO | |
Bovine, Human, Porcine, Rabbit, Sheep | |
IgG2a κ |