missing translation for 'onlineSavingsMsg'
Learn More

Insulin Receptor (α-subunit) Ab-2 Mouse Monoclonal Antibody, Epredia™

Epredia™ Insulin Receptor (α-Subunit) Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

Brand:  Epredia MS-632-P0

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12671837

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.


Description

Description

The human insulin receptor is a heterotetrameric membrane glycoprotein consisting of disulfide-linked subunits in a Β-α-α-Β configuration. The Β-subunit (95kDa) possesses a single transmembrane domain, whereas the α-subunit (135kDa) is completely extracellular.

Host Species: Mouse

Clone: 83-7

Isotype: IgG1/κ

Species Reactivity: Human, Cow, Pig, Sheep and Rabbit. Does not react with mouse and rat.

Epitope: Exon 3

Immunogen: IM-9 lymphocytes followed by IR

Molecular Weight: 135kDa

Positive Control: IM-9 lymphocyte cells, Placenta

Cellular Localization: Cell membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • ELISA
  • Flow Cytometry
  • Insulin-like Agonist
  • Does Not Significantly Inhibit Insulin Binding
  • Tyrosine Kinase Assay (Ab-mediated capture on microplates)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Insulin Receptor (alpha-Subunit) Ab-2
Monoclonal
200μg/mL
Mouse
100 μL
Steroids Hormones and Growth Factors
Bovine, Human, Porcine, Rabbit, Sheep
IgG2a κ
Immunohistochemistry (Paraffin)
83-7
Unconjugated
IM-9 lymphocytes followed by IR
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions