missing translation for 'onlineSavingsMsg'
Learn More

Pan-Cytokeratin Ab-2 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Epredia™ Cytokeratin, Pan Ab-2, Mouse Monoclonal Antibody.

Brand:  Epredia MS-356-P1ABX

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12631157

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.


Description

Description

Twenty human keratins are resolved with two-dimensional gel electrophoresis into acidic (pI <5.7) and basic (pI >6.0) subfamilies. Members of the acidic and basic subfamilies are found together in pairs. The composition of keratin pairs varies with the epithelial cell type, stage of differentiation, cellular growth environment, and disease state. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.

Host Species: Mouse

Clone: PAN-CK

Isotype: IgGs

Species Reactivity: Human. Others not known.

Epitope: Not determined

Positive Control: Skin

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Cytokeratin, Pan Ab-2
Monoclonal
1mg/mL
Mouse
RUO
Primary
Concentrated
Immunofluorescence, Immunohistochemistry (Paraffin)
PAN-CK
Unconjugated
100 μL
Cytoskeletal Protein
Human
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions