missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Permanent Fast-Red Quanto

Epredia™ Lab Vision Permanent Fast-Red Quanto provides a permanent magenta stain for use with immunohistochemistry procedures.

Brand:  Epredia™ TA-125-QAL

Additional Details : Weight : 0.18000kg

 View more versions of this product

Product Code. 12683967

  • 575.00€ / 125mL


Estimated Shipment
25-05-2021
Add to basket

Description

Description

Historically, integrating fast red chromogens into routine Imunohistochemistry was a challenge. Epredia Permanent Fast-Red Quanto eliminates these challenges and produces the brilliant results you expect.
  • Permanent Results — Does not fade during long term storage
  • Solvent Resistance — Insoluable in xylene or alcohols allowing Fast-Red slides to be coverslipped with routine HEs and IHC slides
  • Automated “On Slide” Chromogen Mixing — The two chromogen reagents are delivered in equal parts and are mixed sequentially, directly on the slide, allowing any open instrument to mix the chromogen on board reducing turn around time and manual steps
  • Fast Incubation Times — With typical incubation times of 11 to 15 minutes Permanent Fast Red Quanto is one of the quickest fast reds on the market
Specifications

Specifications

125mL
Store at 2° to 8°C
Videos
SDS
Documents