missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Lab Vision™ Thrombospondin (TSP) Ab-11, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Thrombospondin (TSP) Ab-11, Mouse Monoclonal Antibody.

Brand:  Thermo Scientific™ MS-1066-P

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12628806

  • 775.00€ / 1mL


Estimated Shipment
01-04-2021
Add to basket

Description

Description

Provide accurate, reproducible results with the Thermo Scientific™ Thrombospondin (TSP) Ab-11, Mouse Monoclonal Antibody.

Thrombospondin is a protein from platelet a-granules. It is made up of three identical subunits bound by interchain disulfides. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.

Host Species: Mouse

Clone: D4.6 + A6.1 + MBC 200.1

Isotype: IgG1 + IgG1 + IgG1

Species Reactivity: Human. Others not known.

Immunogen: Purified human TSP

Molecular Weight: ∽450kDa (nonreduced), 170 to 180kDa (reduced)

Cellular Localization: Secretory granules, Golgi complex, endoplasmic reticulum, extracellular matrix

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting (Reduced and nonreduced)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Thrombospondin (TSP) Ab-11
Monoclonal
200μg/mL
Concentrated
Purified human TSP
1mL
Angiogenesis
Primary
Human
Western Blot
D4.6 + A6.1 + MBC 200.1
Unconjugated
Murine
IgG1
RUO
Primary
Monoclonal
Videos
SDS
Documents