Learn More
Epredia™ Lab Vision™ UltraVision™ ONE Detection System: AP Polymer (Ready-To-Use)
Shop All Epredia ProductsDescription
Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Enzyme: Alkaline Phosphatase
Chromogen/Substrate: None provided
Includes:
TL-060-ALJ: 60mL Ultra V Block (TA-060-UB), 60mL UltraVision ONE AP Polymer (TL-060-APJ)
TL-125-ALJ: 125mL Ultra V Block (TA-125-UB), 125mL UltraVision ONE AP Polymer (TL-125-APJ)
Note:
UltraVision ONE AP polymer detection system detects rabbit and mouse immunoglobulins bound to an antigen in tissue sections. The specific primary antibody is located by a universal secondary antibody polymer formulation. The amino acid polymer is conjugated to alkaline phosphatase and the Fab fragments of goat anti-rabbit and goat anti-mouse. The polymer complex is then visualized with an appropriate substrate/chromogen.
Specifications
Specifications
| Chromagen | None provided |
| Count | 300 to 600 slides |
| Enzyme | Alkaline Phosphatase |
| Product Line | Lab Vision, UltraVision |
| Specificity | anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L) |
| Substrate | None provided |
| Includes | 60mL Ultra V Block (TA-060-UB), 60mL UltraVision ONE AP Polymer (TL-060-APJ) |
| Quantity | 60 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.