Learn More
MP Biomedicals LabCare Cell Culture Room Antimicrobial Spray
Shop All MP Biomedicals Products
Click to view available options
Size:
100 mL
450 mL
1.8 L
Unit Size:
1.8 L
100mL
450mL
Description
- MP Biomedicals LabCare Cell Culture Room Antimicrobial Spray is an advanced, broad-spectrum solution formulated to eliminate Viruses, Bacteria, Fungi,
- Mycoplasma, and molds within cell culture rooms and laboratories
- This low-toxicity microbicide and preservative solution effectively eradicates diverse microorganisms known to compromise cell cultures
- Its potent formula ensures lasting protection with properties that endure for 7-10 days, maintaining optimal laboratory hygiene and cell culture integrity
- LabCare Cell Culture Room Antimicrobial Spray is intended for effective and safe elimination of viruses, bacteria, fungi, mycoplasma, and molds within cell culture rooms and laboratories
Specifications
Specifications
| Product Type | Disinfectant Spray |
| Color-coded | No |
| Form | Liquid Spray |
| Scent | Unscented |
| Sterility | Non-sterile |
| Size | 450 mL |