missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals LabCare Water Bath Disinfectant Solution

Product Code. p-200113577 Shop All MP Biomedicals Products
Click to view available options
Size:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 30281214

Brand: MP Biomedicals 093051349

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Special formulated solution for microorganisms removal in water baths.

  • MP Biomedicals LabCare Water Bath Disinfectant Solution is a broad-spectrum, high-efficiency, Stable, low-toxic, antimicrobial microbicide and preservative solution
  • It is specifically formulated for inhibiting and removing microorganisms from water in water baths or incubator tanks
  • Ensures water cleanliness and prevents cross-contamination
TRUSTED_SUSTAINABILITY

Specifications

Product Type Disinfectant Solution
Color-coded No
Form Liquid
Scent Unscented
Sterility Non-sterile
Size 100 mL
compliance-icons compliance-icons
Product Identifier
  • LabCare Water Bath Disinfectant Solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : 1,2-benzisothiazol-3(2H)-one, benzyldodecyldimethylammoniumbromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.