Learn More
Labconco™ Atmospure Adapter Kit
Adapters allow 3/4 in. hose to connect to 3/8 in. fittings on glove boxes
Brand:  Labconco™ 5441900
Additional Details : Weight : 2.30000kg
 View more versions of this product
Product Code. 15967590
  • 319.00€ / Each
Estimated Shipment: 05-07-2024
to see stock.
Description
Description
AtmosPure 3/4 in. to 3/8 in. Adapter Kit has fittings that permit 3/4 in. ID neoprene hose included with 5441800 and 5441801 Kits (sold separately) to attach to glove box’s 3/8 in. inlet/outlets. Two fittings included with each kit. Elbow adapters for use with Labconco Precise Glove Boxes.
Specifications
Specifications
Precise™ Controlled Atmosphere Glove Box
2.3 Kg
Permits 0.75 in. I.D. hose to attach to glove box with 0.37 in. inlet/outlets
5 lb.
Glove Box Kit