Learn More
Labconco™ Atmospure Cable
Product Code. p-4366686 Shop All Labconco Corporation Products
Click to view available options
Product Type:
Atmospure 6-Pin Cable
Unit Size:
Each
Product Code. 15977590
Brand: Labconco™ 5442000
Oxygen/Moisture monitor cable for use with Labconco Protector Glove Boxes
AtmosPure 6-Pin Communication Cable is required for use when AtmosPure Re-Gen Gas Purifiers and glove box configured with Auto Pressure Controller are sharing one vacuum pump.
Specifications
For Use With (Equipment) AtmosPure
Weight (English) 2 lb.
Weight (Metric) 0.9 Kg
Product Type Atmospure 6-Pin Cable
For Use With (Application) For use when Atmospure and glove box with auto pressure controller are sharing one vacuum pump