Learn More
Labconco™ Microcentrifuge Tube Racks
Product Code. 16358887 Shop All Labconco Corporation Products
Click to view available options
Positions:
12
18
96
Holds:
Microtubes (0.5 mL)
Microtubes (1.5 mL)
Microtubes (5 mL)
For Use With (Equipment):
0.5 mL Microtubes
1.5 mL Microtubes
5 mL Microtubes
Dimensions (L x W x H):
5 x 2 x 2.5 in.
6 x 2 x 1.5 in.
8 x 6 x 1.5 in.
8 x 8 x 1.5 in.
Unit Size:
Each
Product Code. 16358887
Brand: Labconco™ 4026401
Only null left
Add to basket
Add to basket
Hold tubes upright and promote conduction of heat or cold from Temp-Zone area on selected models of Purifier Cell Logic Biosafety Cabinets to help maintain sample temperature
  • Material: Anodized aluminum
  • Color: Blue
Specifications
Product Type Microcentrifuge Tube Rack
Array 96
Autoclavable Autoclavable
Color Natural
Holds Microtubes (0.5 mL)
Material Aluminum
Positions 96
Shape Rectangular
Shelves None
Stackable No
Sterility Non-sterile
For Use With (Equipment) 0.5 mL Microtubes
Dimensions (L x W x H) 8 x 6 x 1.5 in.
Length (Metric) 20.3 cm
Length (English) 8 in.
Width (Metric) 15.2 cm
Width (English) 6 in.
Height (Metric) 3.8 cm
Height (English) 1.5 in.