Learn More
Labconco™ Replacement Filters for Benchtop Fume Adsorbers
Replacement filters for use with Labconco Fume Adsorbers and Fume Adsorber-2s.
620.00€
Specifications
Filter Type | Carbon |
---|---|
For Use With (Equipment) | Benchtop Fume Adsorber |
Weight (English) | 11 lb. |
Weight (Metric) | 5 Kg |
Product Type | Filter |
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | |||||
15332822
|
Labconco™
6961300 |
Organic Vapor |
620.00€
Each |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15312822
|
Labconco™
6961301 |
Formaldehyde-formalin |
620.00€
Each |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Replacement Ammonia Carbon Filter for use with ammonia and low molecular weight amines. 8.8 pounds of impregnated carbon. Adsorption weight 10% (0.88 lbs.). Adsorption volume 800 ml of 50% ammonia. For use with Labconco Fume Adsorber and Fume Adsorber-2. Fume Adsorber Models 3955202 and 3955222 include one filter; Fume Adsorber-2 Models 3955402 and 3955422 include two filters.Specifications
Carbon | |
11 lb. | |
Filter |
Benchtop Fume Adsorber | |
5 Kg |