Learn More
Labconco™ Replacement Prefilter for Protector Downdraft Powder Station
Replacement Prefilter for use in Protector Downdraft Powder Stations.
Brand:  Labconco™ 3768901
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 15470412
  • 31.05€ / Each
Estimated Shipment: 19-07-2024
to see stock.
Description
Description
Replacement Prefilter for use in Protector Downdraft Powder Stations.
Replacement Prefilter for use with 4-Ft, 5-Ft, 6-Ft and 8-Ft Purifier Horizontal Clean Benches. Two prefilters required for 4-Ft, one required for 5-Ft, three required for 6-Ft and four required for 8-Ft. Labconco recommends replacing prefilters every three months.
Specifications
Specifications
Prefilter
1 in.
0.5 Kg
23.8 in.
2.5 cm
60.3 cm
45.1 cm
Particulate
4 ft. Purifier™ horizontal clean benches and 6 ft. Purifier™ horizontal clean benches
1 lb.
20 to 30 in.
17.8 in.
20 to 30 x 16.5 x 1 in. (50.8 to 76.2 x 41.9 x 2.5 cm)
50.8 to 76.2 cm
Prefilter