Learn More
Labconco™ ScrubAir™ Pipette Washer / Dryer Accessory, Spindle Kit
Easily adapt your ScrubAir glassware washer / dryer to permit cleaning of small test tubes or vials with the Labconco ScrubAir Spindle Kit.
Brand: Labconco™ 4615700
Additional Details : Weight : 0.50000kg
Description
- Outfits small test tubes or vials, 7 mm ID or larger, to be washed in the ScrubAir
- Kit contains 24 each 5mm OD polystyrene plastic spindles with caps and stainless steel test tube rack to hold up to 20 each test tubes
- Shipping weight: 0.9 kg (2.0 lbs.)
Specifications
Kit | |
ScrubAir Pipette Washer/Dryer |