missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Single Electrical Receptacle Kits: International GFCI Switch
Mount on the left or right side of any 3, 4, 5, 6, 7 or 8 ft. Protector Premier, XStream, XL, PVC Perchloric Acid or Stainless Steel Laboratory Hood
Brand: Labconco™ 9414100
Additional Details : Weight : 0.90719kg
Includes: Electrical receptacle, wiring, junction box and receptacle cover plate
Specifications
Protector™ Premier™, XStream™, XL™ fume hood | |
4 lb. | |
9 in. | |
22.9 cm |
2 in. | |
1.8 Kg | |
5.1 cm | |
International GFCI switch |