missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ SteamScrubber™ Labware Inserts

Hold a variety of wider-mouth flasks, beakers and other labware in inverted position

Brand:  Labconco™ 4401901

Additional Details : Weight : 0.45000kg

 View more versions of this product

Product Code. 15109665

  • 77.90€ / Each


Estimated Shipment
04-06-2020
Add to basket

Description

Description

  • Pins have plastic tips to protect glassware from scratching
  • Retainer Top is recommended for use with plastic and lightweight glassware
  • Type 304 stainless steel
  • Fits standard bottom rack on SteamScrubber units only

Specifications

Specifications

20.32cm
Labware insert of FlaskScrubber™, SteamScrubber™ glassware washer
0.7cm
0.7kg
Each
Type 304 Stainless Steel
Retainer Top
46.9cm
Videos
Documents