missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Utility Shelf Kit
Keeps accessories in reach
Brand: Labconco™ 3925000
Additional Details : Weight : 2.30000kg
Description
Keeps accessories in reach
Three one-piece, powder-coated steel shelves attach to rear baffle perforations.
- Printer shelf (can also be inverted and rest atop enclosure); 6.75W × 10 in.D (17.1 × 25.4cm)
- Utensil shelf with scalloped front edge supports spatulas, brushes and other items; 5.25W × 9 in.D (13.3 × 22.9cm)
- Miscellaneous shelf, 5.25W × 7 in.D (13.3 × 17.8cm)
- Fits XPert™ Balance Enclosure, Station or System; Protector™ XVS Ventilation Station; Purifier™ Class I and HEPA-Filtered Enclosures (manufactured after July 2004); and Protector Work Station (manufactured after August 2004)
Specifications
5 lb. | |
2.3 Kg | |
Utility Shelf Kit |