Learn More
Laboratory Performance Check Standard without Mercury For Environmental Analysis, SPEX CertiPrep™
Shop All SPEX Certiprep Products
Click to view available options
Quantity:
125 mL
500 mL
Unit Size:
125mL
500mL
Description
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| K | 100μg/mL |
| P | 100μg/mL |
| SiO&subscript_2; | 100μg/mL |
| Al | 20μg/mL |
| As | 20μg/mL |
| B | 20μg/mL |
| Ba | 20μg/mL |
| Be | 20μg/mL |
| Ca | 20μg/mL |
| Cd | 20μg/mL |
| Co | 20μg/mL |
| Cr | 20μg/mL |
| Cu | 20μg/mL |
| Fe | 20μg/mL |
| Li | 20μg/mL |
| Mg | 20μg/mL |
| Mn | 20μg/mL |
| Mo | 20μg/mL |
| Na | 20μg/mL |
| Ni | 20μg/mL |
| Pb | 20μg/mL |
| Sb | 20μg/mL |
| Se | 20μg/mL |
| Sn | 20μg/mL |
| Sr | 20μg/mL |
| Tl | 20μg/mL |
| V | 20μg/mL |
| Zn | 20μg/mL |
| Ag | 5μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Chemical Name or Material | LPC Standard 1 without Mercury |
| Packaging | Triple-leached LDPE bottle |
| Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
| Technique | AA, ICP |
| For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
| Type | Certified Reference Material |
| Solvent or Matrix | Nitric Acid/Trace Tartaric Acid/Trace Hydrogen Fluoride |
| Quantity | 125 mL |
| Grade | Environmental |
| Concentration or Composition (by Analyte or Components) | Various |
Safety and Handling
Product Identifier
- LPC Standard 1
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P363-Wash contaminated clothing before reuse.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : nitric acid, hydrofluoric acid
Product intended for laboratory use only