Learn More
Thermo Scientific™ Laboratory Refrigerator and Freezer Casters
Achieve higher elevation on high-performance refrigerators and freezers with these laboratory refrigerator and freezer casters. Casters offer a 2 or 3 in. height increase.
Brand:  Thermo Scientific™ 6041-1
Additional Details : Weight : 5.00000kg
 View more versions of this product
Product Code. 15752436
  • 368.00€ / Pack of 4
Estimated Shipment: 02-09-2024
to see stock.
Add to basket
Description
Description
Achieve higher elevation on high-performance refrigerators and freezers with Thermo Scientific™ Laboratory Refrigerator and Freezer Casters. Casters offer a 2 or 3 in. height increase for Thermo Scientific™ Revco™ , Forma™ and Puffer Hubbard™ laboratory refrigerators and freezers.
  • An option for replacement on most models
  • Factory-installed when ordered with unit; customer-installed if ordered separately
  • 2 in. (50mm) casters are standard on high-performance refrigerators and freezers
  • Optional 3 in. (76mm) casters are shipped with the cabinet but must be installed upon delivery
Ordering Information:
  • 6041: 2 in. casters (50mm), set of four
  • 6041-1: 3 in. casters (76mm), set of four
  • 6983: Locking casters for -30°C freezers only, includes two nonlocking 2 in. (50mm) casters
Specifications
Specifications
3 in. (7.6cm) Dual Wheel
78.8 cu. ft.
-30°C
Caster
2231.3 L
4.9, 11.5, 23.3, 29.2, 51.1, 78.8 cu. ft. Units