Learn More
BOLA Laboratory Screw Joints with O-ring
Product Code. 10565742
Click to view available options
Diameter (Metric):
0.8 mm
1.6 mm
2 mm
Unit Size:
Each
Product Code. 10565742
Brand: BOLA D59302
Only null left
Add to basket
Add to basket
Laboratory Screw Joints with O-ring Glass-fibre reinforced ETFE cap, inner parts made of PTFE and PVDF, with O-ring
Red screw cap made of glass-fibre reinforced ETFE, inner parts consisting of a v-ring (ETFE), a tapered ring and a sealing ring (both PTFE) as well as an o-ring (FKM) for tubing diameters under 3 mm, (not exposed to the medium). Very good chemical resistance, the medium is only exposed to PTFE.
Specifications
Certifications/Compliance FDA
Diameter (Metric) 0.8 mm
Autoclavable Autoclavable
Thread Size 14
Color White
Flexibility Rigid