Learn More
Labtainer Labtainer w/Quick Connects BPC 2L Aegis5-14
GREENER_CHOICE
Brand:  Labtainer SH30963.05
Labtainer w/Quick Connects BPC 2L Aegis5-14
Product Code. 17835726
  • ONLINE EXCLUSIVE PRICE
    895.00€
    738.15€ / Pack of 10
    Save 156.85€
    17% Off