missing translation for 'onlineSavingsMsg'
Learn More

Karl Hecht™ Labtray 2

Product Code. 10320841
Click to view available options
Dimensions (L x W):
35 x 25 cm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10320841

Brand: Karl Hecht™ 42002008

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Designed in consideration of the photometric hemoglobin determination.

  • Plastic tray
  • Plexiglas™ test tube rack for 24 tubes
  • Plexiglas stand with 3 compartments and 3-place bottle holder
  • 3 amber glass bottles with frosted label for inscription, two wide neck, one narrow
  • Test tubes of 100 x 16mm are recommended
  • Free space on tray is intended for filled pipettes and blood smears

Specifications

Dimensions (L x W) 35 x 25 cm
Color White
Type Laboratory Tray
For Use With Photometric hemoglobin determination
Includes Glass Bottles, Stand, Test Tube Rack
Shape Rectangular
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.