Learn More
Brady™ Laminated Polyester Radiation Laser Sign
Product Code. p-8001110
Click to view available options
Displayed Text:
Laser beam warning
Warning of radioactive materials or radiation ioniserenden
Unit Size:
Each
Product Code. 11760194
Brand: Brady™ 223479
Only null left
Specifications
Alternate Part Number PIC 304-TRI 315-B7541
Material Polyester
Quantity 1 Ea.
Adhesive Type Permanent Acrylic
Description Laminated Polyester
For Use With (Application) Pipe markers and safety signs
Height (Metric) 315 mm
Max. Temperature (Metric) 110°C
Thickness 0.024 mm
Width (Metric) 315 mm
Shape Triangular
Displayed Text Warning of radioactive materials or radiation ioniserenden
Product Type Radiation Laser Sign
Color Black on Yellow