Learn More
Brady™ Laminated Polyester Stickers for the Transport of Infectious Substances
Product Code. 10755145
Change view
Click to view available options
Height (Metric):
100 mm
200 mm
297 mm
Width (Metric):
100.00 mm
200.00 mm
297.00 mm
Unit Size:
Each
Product Code. Height (Metric) Width (Metric) unitSize
10755145 100 mm 100.00 mm Each
11744884 297 mm 297.00 mm Each
10080284 200 mm 200.00 mm Each
3 options
Product Code. 10755145
Brand: Brady™ 257534
Only null left
Specifications
Alternate Part Number ADR 6.2-100*100-B7541
Finish Gloss
Material Laminated Polyester
Quantity 1 Ea.
Adhesive Type Permanent Acrylic
Height (Metric) 100 mm
Thickness 0.099 mm
Width (Metric) 100.00 mm
Temperature Range (Metric) -40 °C-110 °C
Shape Rhomboid
Displayed Text 6
Color Black on White