Learn More
Brady™ Laminated Radioactive Polyester Warning Sign
Click to view available options
Unit Size:
Each
Specifications
Specifications
| Alternate Part Number | PIC 304-TRI 050-B7541 |
| Material | Polyester |
| Quantity | 1 Ea. |
| Adhesive Type | Permanent Acrylic |
| Description | Laminated Polyester |
| For Use With (Application) | Pipe markers and safety signs |
| Height (Metric) | 50 mm |
| Max. Temperature (Metric) | 110°C |
| Thickness | 0.024 mm |
| Width (Metric) | 50 mm |
| Show More |