Description
- No reagents or field kits required for most strips
- Chemically treated pads on plastic strips produce color change
- Pop-top vials have molded desiccant liner and hinged snap cap to protect from moisture
- As little as 10-second development time
Test strips for testing water: wastewater, industrial water boiling and cooling, drinking water, laundry and sanitation †
†Strips have been evaluated for use in these applications. Use in other applications is subject to potential interferences.
Specifications
Specifications
| For Use With (Application) | Water quality testing |
| Parameter Specifications | Peracetic acid |
| Quantity | 50 Strips/Vial |
| Range | 0-10-20-40-60-85-160 ppm |
| Storage Requirements | Store tightly sealed vial in a cool, dry storage area away from moisture; Keep out of direct sunlight |
| Type | Single Factor Test Strip |