Learn More
Lanthanum(III) iodide, ultra dry, 99.99% (metals basis), Thermo Scientific™
74.30€ - 250.00€
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15427455
|
Thermo Scientific Alfa Aesar
047184.03 |
1g |
75.80€ 74.30€ / 1g Save 1.50€ 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15437455
|
Thermo Scientific Alfa Aesar
047184.06 |
5g |
255.00€ 250.00€ / 5g Save 5.00€ 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Ampouled under argon
Chemical Identifiers
| 13813-22-4 | |
| MFCD00049470 |
Specifications
| 13813-22-4 | |
| Soluble in dimethyl form amide. (578.7 g dm-3 at 20°C). Soluble in water. | |
| 99.99% | |
| Ampule | |
| (metals basis) |
| MFCD00049470 | |
| 519.62 | |
| Odorless | |
| Light and moisture sensitive | |
| Lanthanum(III) iodide, Ultra dry |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 237-465-6
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Ampouled under argon
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.