missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lanthanum solution for ICP/MS 10mg/L HNO3 matrix, Fisher chemical™
Click to view available options
Quantity:
50 mL
100 mL
Packaging:
Bottle
Specifications
Concentration | 10mg/L |
Packaging | Bottle |
Quantity | 100 mL |
Solvent or Matrix | HNO3 |
Chemical Name or Material | Lanthanum |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'