Learn More
Invitrogen™ LanthaScreen™ Amine Reactive Tb Chelate
LanthaScreen™ Amine Reactive Tb Chelate
Brand:  Invitrogen™ PV3581
Additional Details : Poids : 0.05250kg
 View more versions of this product
Product Code. 10595343
  • 13690.00€ / 1mg
Expédition estimée
24-12-2020
Add to basket
Description
Description
As part of the LanthaScreen™ TR-FRET toolbox of assay reagents, LanthaScreen™ Amine Reactive Tb Chelate labeling reagent is available for assay development. The amine-reactive, isothiocyanate group conjugates to virtually any peptide or protein that contains one or more accessible amine moieties, such as the amine group found on a lysine residue or an unmodified N-terminal amine group of a peptide or protein. Amine-modified DNA oligonucleotides can be labeled as well. Energy transfer from the terbium donor to a suitable acceptor such as fluorescein is readily detected by monitoring an increase in acceptor fluorescence intensity. See the user guide for application-based protocols for labeling peptides containing free amino groups, amine-modified oligonucleotides, and IgG antibodies.
Specifications
Specifications
LanthaScreen™ Amine Reactive Tb Chelate, 1mg, 1050.88g/mol Molecular Weight, ≥90% by HPLC Purity, Store at –20°C, Used For Pharma and Biopharma, Target and Lead Identification and Validation
LanthaScreen™ TR-FRET labeling reagents are provided lyophilized
LanthaScreen™ Amine Reactive Tb Chelate
1mg
≥90% by HPLC
Drug discovery & development, target & lead identification & validation, TR-FRET assay development
LanthaScreen™ TR-FRET labeling reagents are provided lyophilized
–20°C
1050.88g/mol