missing translation for 'onlineSavingsMsg'
Learn More

LanthaScreen™ Tb-p4E-BP1 (pThr46) Antibody Kit

LanthaScreen™ Tb-p4E-BP1 (pThr46) Antibody Kit

3920.00€ - 18840.00€

Specifications

Antigen Serine/threonine kinase
Concentration 0.52mg/mL
Applications Kinase Assay, Enzyme and Protein Activity Assays, Kinase Assay
Conjugate Tb (Terbium)/GFP (EmGFP)/Lanthanide Chelates
Regulatory Status RUO, RUO
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10053143
View Documents Promotion Details
Invitrogen
PV4757
25 μg
3920.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 16-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
10793484
View Documents Promotion Details
Invitrogen
PV4758
1 mg
18840.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 16-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

A part of the PI3K/AKT Pathway, 4E-BP1 (EIF4EBP1; eukaryotic translation initiation factor 4E binding protein 1) is a cytosolic protein that is phosphorylated at Thr46 by mTORC1 upon stimulation of the pathway.

For use in combination with the GFP-4E-BP1 physiological protein substrate (PV4759) to evaluate inhibitors in medium to high throughput screening applications. An optimized mTor assay buffer (PV4794) is also available.
Specifications

Specifications

Serine/threonine kinase
Kinase Assay, Enzyme and Protein Activity Assays, Kinase Assay
RUO, RUO
Primary
150kDa
0.52mg/mL
Tb (Terbium)/GFP (EmGFP)/Lanthanide Chelates
Target and Lead Identification and Validation
–20°C
1 Kit(s)
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.