Learn More
Thermo Scientific™ LanthaScreen™ Thiol Reactive Tb Chelate
Product Code. 10082552
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
Unit Size:
100µg
10µg
1mg
Product Code. Quantity unitSize
10082552 1 mg 1mg
11543467 10 μg 10µg
11533467 100 μg 100µg
3 options
Product Code. 10082552
Brand: Thermo Scientific™ PV3578
LanthaScreen™ Thiol Reactive Tb Chelate
  • As part of the LanthaScreen™ TR-FRET toolbox of assay reagents, LanthaScreen™ Thiol Reactive Tb Chelate labeling reagent is available for assay development
  • The thiol-reactive maleimide group can be conjugated to virtually any peptide or protein that contains one or more accessible thiol moieties, such as the thiol group found on a cysteine residue
  • Energy transfer from the terbium donor to a suitable acceptor such as fluorescein is readily detected by monitoring an increase in acceptor fluorescence intensity
  • See the user guide for an application-based protocol for cysteine-containing peptides or proteins
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 mg
Product Type Thiol Reactive Tb Chelate
Content And Storage LanthaScreen™ TR-FRET Labeling Reagents are provided lyophilized. Store at -20°C, desiccated, and protected from light. After reconstitution, terbium lanthanide chelates should be stored on ice and used within the same day.
Shipping Condition Dry Ice
Product Line LanthaScreen