Learn More
Brady™ LASERTAB™ Laser Printable Labels, B-747 Polyester
Product Code. 11583564
Click to view available options
Size (English):
0.25 x 0.25 in. (W x H)
0.3 x 0.25 in. (W x H)
0.375 x 0.375 in. (W x H)
0.5 x 0.437 in. (W x H)
0.65 x 0.2 in. (W x H)
0.65 x 0.25 in. (W x H)
0.8 x 0.25 in. (W x H)
0.8 x 0.5 in. (W x H)
1 x 0.5 in. (W x H)
1.5 x 0.25 in. (W x H)
1.875 x 0.375 in. (W x H)
1.875 x 0.833 in. (W x H)
1.875 x 1.33 in. (W x H)
2.5 x 1 in. (W x H)
3 x 0.375 in. (W x H)
8.5 x 11 in. (W x H)
Color:
White
Yellow
Unit Size:
1 pack
Product Code. 11583564
Brand: Brady™ 029700
Only null left
Add to basket
Add to basket
Designed for use with laser printers. Brady LASERTAB Laser Printable Labels are made of B-747 polyester.
  • Blank, printable polyester labels for Laser Printers
  • CSA Accepted material when printed with designated laser printer toners
Specifications
Certifications/Compliance UL Recognised; CSA Accepted
Description Chemical, high temperature, humidity, smear, UV and weather-Resistant
Adhesive Type Acrylic, Permanent
Coating None
Color White
Finish Matte
Format Die-cut
Material Polyester (B-747)
Printable Sides Single-side Printable
Print Technology Laser
Product Type Laser Printable Label
Sterility Non-sterile
For Use With (Application) Circuit board labelling, General identification
For Use With (Equipment) Laser Printers
Horizontal Repeat (Metric) 17.78 mm
Labels per Row 22
Min. Operating Temperature (Metric) -40°C
No. per Pack 10032 Labels/Pack
Quantity 10,000/Pk.
Shape Rectangle
Size (English) 0.3 x 0.25 in. (W x H)
Size (Metric) 7.62 x 6.35 mm (W x H)
Temperature Range (English) -40°F to +266°F
Temperature Range (Metric) -40°C to +130°C
Thickness (English) 0.0037 in.
Thickness (Metric) 0.094 mm
Dimensions (LxW) 6.35 x 7.62 mm
Length (English) 0.3 in.
Width (English) 0.3 in.
Width (Metric) 7.62 mm
Width (Metric) Printable Area 7.62 mm
Height (English) 0.25 in.
Height (English) Printable Area 0.25 in.
Height (Metric) 6.35 mm
Height (Metric) Printable Area 6.35 mm
Alternate Part Number LAT-1-747-10