Learn More
Kolmi Hopen™ Latex-free Clip Caps in Wall Dispenser, 10 g/m2
GREENER_CHOICE
Product Code. p-7227138 Shop All Medicom Products
Click to view available options
Color:
Blue
White
Includes:
Caps and Wall Dispenser
Clip Caps and Polybags
Unit Size:
Pack of 150
Pack of 300
Product Code. 17710149
Brand: Kolmi Hopen™ C12004
Only null left
Add to basket
Add to basket
Elasticated polypropylene clip caps for use in medical and industrial settings.
  • 100% polypropylene spunbonded, non-woven
  • Bonding and ultrasonic assembly
  • Synthetic elastic under hem for better comfort
  • Soft, very resistant and breathable for better comfort
  • Great stretch to wrap and hold all hair
  • 10 g/m2 fabric weight
  • Made in France
Specifications
Material Polypropylene
Color White
Disposable Single-use
Design Type Clip Cap
Latex Free Yes
Certifications/Compliance CE, EN 14683 Type: I
Quantity 300 Pk.
Includes Caps and Wall Dispenser
For Use With (Application) Medical, Industrial, Laboratory