missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ LB Broth, Lennox (Pre-Buffered Capsules)
Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.
177.00€ - 187.00€
Specifications
Format | Pre-Buffered Capsules |
---|---|
Product Type | Microbiology Media |
Formulation | Casein Peptone 10g/L, Yeast extract 5g/L, Sodium Chloride 5g/L, TRIS/TRIS HCl 1.5g/L |
For Use With (Application) | For maintenance and propagation of Escherichia coli in molecular biology procedures |
Product Code | Brand | Quantity | Packaging Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging Type | Price | Quantity & Availability | ||||
11326002
|
Fisher BioReagents™
BP9730-500 |
25 x 20 g | Poly Bottle |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
11356002
|
Fisher BioReagents™
BP9733-500 |
500 x 1 g | Poly Bottle |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
Description
- Directions for Media Preparation: Add 1 capsule per 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water). Heat to completely dissolve capsule. Before use, sterilize by autoclaving for 15 minutes.
- Product is in capsule form
- Each capsule contains 1g of broth
- Capsules are prebuffered and set to pH 7.2 at 20°C using a 10mM solution of tris/tris HCl
- pH will vary with temperature
Specifications
Pre-Buffered Capsules | |
Microbiology Media | |
Casein Peptone 10g/L, Yeast extract 5g/L, Sodium Chloride 5g/L, TRIS/TRIS HCl 1.5g/L | |
For maintenance and propagation of Escherichia coli in molecular biology procedures |