missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ LB Broth, Miller (Pre-Buffered Capsules)

Product Code. 11346002
Click to view available options
Quantity:
25 x 25 g
500 x 1 g
Packaging Type:
Poly Bottle
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 11346002

Brand: Fisher BioReagents™ BP9732500

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.

  • Directions for Media Preparation: Add 1 capsule per 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water). Heat to completely dissolve capsule. Before use, sterilize by autoclaving for 15 minutes.
  • Product is in capsule form
  • Each capsule contains 1g of broth
  • Capsules are prebuffered and set to pH 7.2 at 20°C using a 10mM solution of tris/tris HCl
  • pH will vary with temperature
TRUSTED_SUSTAINABILITY

Specifications

Format Pre-Buffered Capsules
Media Type LB Broth
Product Type Microbiology Media
Formulation Casein Peptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L, TRIS/TRIS HCl 1.5g/L
For Use With (Application) For maintenance and propagation of Escherichia coli in molecular biology procedures
Packaging Type Poly Bottle
Quantity 500 x 1 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.