missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ LB Medium, Large Capsules

Product Code. 11323239 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
227 g
454 g
1000 g
5000 g
15 kg
Unit Size:
15kg
1kg
227g
454g
5kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11323239 15 kg 15kg
11363229 1000 g 1kg
11393229 5000 g 5kg
11393219 227 g 227g
11333229 454 g 454g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11323239 Supplier MP Biomedicals™ Supplier No. 113002056

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Culture media used for the growth of bacteria.

  • Formulations for the E. coli growth media contain 10g of tryptone and 5g of yeast extract per liter, 10g NaCl per liter
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular Biology Certified™
Content And Storage 15°C to 30°C
Format Capsule
Formulation Tryptone 10g/L, Yeast extract 5g/L, NaCl 10g/L
For Use With (Application) For growth of bacteria
Quantity 15 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.