Learn More
Lovibond™ LC 100 Hand-Held Imaging Spectrophotometer
Product Code. p-4587714
Click to view available options
For Use With (Application):
Food, Bev, Cosmetics, Pharm
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 15564283
Brand: Lovibond™ 401100
Regulatory: This product is under eco-contribution tax.
Creates conformance reports in either PDF or Excel formats that can easily be shared with PC or compatible printer
  • Each measurement combines 27 images illuminated with different colours and from different directions
  • Enables preview of measurement area and storage and recall of image after measurement
  • Measures and stores up to 20 reference standards with each date and time stamped with option to name
  • Stores up to 350 sample measurements on device and compares to reference standards
  • Samples can be tagged via on-screen keyboard for text or via built in voice recorder
  • For indoor use only
LC 100 and SV 100 Integrated Kit
  • Contents: spectrophotometer, holder, sample cells, USB cord, carrying case
  • Food and Beverages, Textile, Chemicals, Pharmaceutical and Cosmetics, Medical and Clinical
Specifications
Display TFT
For Use With (Application) Food, Bev, Cosmetics, Pharm
Includes Spectrophotometer, holder, sample cells, USB cord, carrying case
Measurement Time 1.8 sec.