Learn More
Trajan Scientific and Medical™ LC PEEKsil™ Tubing
Click to view available options
Overall Length (Metric):
50 mm
100 mm
150 mm
200 mm
500 mm
Diameter (Metric) Inner:
0.025 mm
0.05 mm
0.075 mm
0.1 mm
0.15 mm
0.175 mm
0.2 mm
0.3 mm
0.53 mm
Unit Size:
Pack of 2
Pack of 5
Description
- Fused silica internal wall provides smooth surface, free of imperfections
- PEEKsil compatible with most organic solvents and resistant to strong acids
- Effective pH range of 0 to 10
- Flexibility ensures easy connection between columns, detectors and injectors
Specifications
Specifications
| Type | Tubing |
| Includes | 2/Pk. |
| Diameter (English) Outer | 1/16 in. |
| Diameter (Metric) Inner | 0.53 mm |
| Diameter (Metric) Outer | 1.6 mm |
| Overall Length (Metric) | 500 mm |
| Material | Polymer-sheathed Fused Silica |