Learn More
Hach™ LCK Cuvette Tests
Product Code. p-7115587
Click to view available options
Analytes:
Acid Capacity (KS4.3/CaCO3)
Total Organic Carbon
Unit Size:
Each
Pack of 25
Product Code. 10122744
Brand: Hach™ LCK362
Insert, read and finish. Hach™ LCK Cuvette Tests are pre-dosed reagents packaged in a fully equipped box. They provide maximum safety, ease of use and photometric evaluation, while using a minimum amount of reagent/chemicals for an environmentally friendly solution.
Hach LCK Cuvette Tests are officially approved for legally required consent limits. With the help of Addista standard solutions and round robin test solutions, they provide the assurance that analytical results are always correct. The photometer identifies the cuvette test from its barcode (IBR+) and calculates the result immediately. Cuvettes provide precision and safety using the Dosicap Zip that eliminated contact with reagents.
  • Pre-dosed, ready-to-use reagents with no rinsing steps needed
  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system and low amounts of reagents
  • Convenient and error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour-coding
Specifications
Analytes Acid Capacity (KS4.3/CaCO3)
Color Blue
Format Pre-dosed Tubes
For Use With (Application) Water quality testing
For Use With (Equipment) Photometer (DR3900, DR6000, DR1900, DR2800, DR3800, DR5000), Addista standard solutions
Includes Fully equipped box, reagent set
No. of Tests 25
Range 0.5 to 8.0mmol/L KS4.3 / 25 to 400mg/L CaCO3
Storage Requirements 15°C to 25°C
Test Method Hach Method
Type LCK Cuvette Test